Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031388.2 | NP_113565.2 | 1 - 2 | 162 | 106 | |
GenBank mRNA | AK082829.1 | - | 1 - 2 | 89 | 106 | |
AK133417.1 | - | 1 - 2 | 166 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Mus musculus 1700 Probe spans exons |