Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172650.2 | NP_766238.1 | 3 - 4 | 409 | 74 | |
GenBank mRNA | AK033491.1 | - | 3 - 4 | 410 | 74 | |
AK034380.1 | - | 3 - 4 | 409 | 74 | ||
AK049362.1 | - | 3 - 4 | 407 | 74 | ||
AK087865.1 | - | 3 - 4 | 410 | 74 | ||
AK139700.1 | - | 3 - 4 | 409 | 74 | ||
BC061458.1 | - | 3 - 4 | 397 | 74 | ||
BC079581.1 | - | 3 - 4 | 396 | 5702630 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |