Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111324.2 | NP_001104794.1 | 1 - 2 | 201 | 68 | |
GenBank mRNA | AF009366.1 | - | 1 - 2 | 137 | 68 | |
AK030985.1 | - | 1 - 2 | 135 | 68 | ||
AK083374.1 | - | 1 - 2 | 129 | 68 | ||
BB842569.1 | - | 1 - 2 | 162 | 68 | ||
BC053713.1 | - | 1 - 2 | 134 | 30077589 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |