Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177320.2 | NP_796294.2 | 12 - 13 | 2037 | 115 | |
GenBank mRNA | AK089483.1 | - | 12 - 13 | 2042 | 115 | |
AK149847.1 | - | 12 - 13 | 2005 | 115 | ||
AK154643.1 | - | 12 - 13 | 2016 | 115 | ||
AK161050.1 | - | 3 - 4 | 945 | 115 | ||
AK171279.1 | - | 11 - 12 | 2162 | 115 | ||
BC128076.1 | - | 11 - 12 | 1886 | 40060132 | 115 | |
BC128077.1 | - | 11 - 12 | 1886 | 40060133 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |