Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081006.1 | NP_001074475.1 | 5 - 6 | 997 | 119 | |
NM_001177630.2 | NP_001171101.2 | 4 - 5 | 1095 | 119 | ||
NM_001177631.1 | NP_001171102.1 | 4 - 5 | 1095 | 119 | ||
NM_029895.4 | NP_084171.2 | 5 - 6 | 1308 | 119 | ||
NM_178059.5 | NP_835160.2 | 5 - 6 | 1308 | 119 | ||
GenBank mRNA | AB093289.2 | - | 4 - 5 | 885 | 119 | |
AB125594.2 | - | 5 - 6 | 1308 | 119 | ||
AB125595.2 | - | 5 - 6 | 1308 | 119 | ||
AK052020.1 | - | 6 - 7 | 1410 | 119 | ||
AK147510.1 | - | 4 - 5 | 1002 | 119 | ||
BC158051.1 | - | 4 - 5 | 817 | 9007405 | 119 | |
BC172143.1 | - | 4 - 5 | 815 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |