Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285459.1 | NP_001272388.1 | 2 - 3 | 1712 | 88 | |
NM_010230.2 | NP_034360.2 | 4 - 5 | 2382 | 88 | ||
GenBank mRNA | X53599.1 | - | 4 - 5 | 2336 | 88 | |
X62379.1 | - | 2 - 3 | 1712 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |