Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029638.2 | NP_083914.2 | 1 - 2 | 40 | 61 | |
GenBank mRNA | AK144567.1 | - | 1 - 2 | 38 | 61 | |
BC021880.1 | - | 1 - 2 | 41 | 4216952 | 61 | |
BY131896.1 | - | 1 - 2 | 40 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |