Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Siglec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011426.3 | NP_035556.3 | 6 - 7 | 2202 | 64 | |
GenBank mRNA | BC141335.1 | - | 6 - 7 | 1796 | 9055947 | 64 |
BC141336.1 | - | 6 - 7 | 1792 | 9055948 | 64 | |
Z36233.1 | - | 6 - 7 | 2200 | 64 | ||
Z36293.1 | - | 6 - 7 | 2200 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |