Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113569.1 | NP_001107041.1 | 16 - 17 | 1601 | 68 | |
NM_009295.2 | NP_033321.2 | 16 - 17 | 1596 | 68 | ||
GenBank mRNA | AF326545.1 | - | 16 - 17 | 1455 | 68 | |
AF326563.1 | - | 16 - 17 | 1455 | 68 | ||
AK004780.1 | - | 11 - 12 | 1320 | 68 | ||
BC031728.1 | - | 16 - 17 | 1528 | 4480978 | 68 | |
D45903.1 | - | 16 - 17 | 1490 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |