Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290558.1 | NP_001277487.1 | 1 - 2 | 270 | 64 | |
GenBank mRNA | BC019978.1 | - | 1 - 2 | 209 | 4239441 | 64 |
BC022114.1 | - | 1 - 2 | 207 | 5098924 | 64 | |
BY772900.1 | - | 1 - 2 | 271 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |