Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198872.1 | NP_001185801.1 | 7 - 8 | 595 | 129 | |
NM_001198873.1 | NP_001185802.1 | 7 - 8 | 580 | 129 | ||
NM_001198874.1 | NP_001185803.1 | 5 - 6 | 526 | 129 | ||
NM_001198877.1 | NP_001185806.1 | 7 - 8 | 584 | 129 | ||
NM_001198878.1 | NP_001185807.1 | 7 - 8 | 569 | 129 | ||
GenBank mRNA | AK141837.1 | - | 5 - 6 | 494 | 129 | |
AK164322.1 | - | 5 - 6 | 450 | 129 | ||
GU992208.1 | - | 7 - 8 | 486 | 129 | ||
GU992209.1 | - | 7 - 8 | 471 | 129 | ||
GU992211.1 | - | 7 - 8 | 524 | 129 | ||
GU992212.1 | - | 7 - 8 | 509 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |