Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099288.1 | NP_001092758.1 | 7 - 8 | 904 | 64 | |
NM_175003.3 | NP_778168.2 | 7 - 8 | 904 | 64 | ||
GenBank mRNA | AK079081.1 | - | 2 - 3 | 205 | 64 | |
BC056366.1 | - | 7 - 8 | 904 | 6825221 | 64 | |
BC059911.1 | - | 7 - 8 | 904 | 6825221 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |