Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tvp23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026210.4 | NP_080486.1 | 2 - 3 | 194 | 75 | |
GenBank mRNA | AK007707.1 | - | 2 - 3 | 110 | 75 | |
AK017572.1 | - | 2 - 3 | 110 | 75 | ||
AK050027.1 | - | 2 - 3 | 230 | 75 | ||
AK077277.1 | - | 2 - 3 | 196 | 75 | ||
AK150936.1 | - | 2 - 3 | 186 | 75 | ||
BC115504.1 | - | 2 - 3 | 174 | 40101013 | 75 | |
BC115505.1 | - | 2 - 3 | 174 | 40101015 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |