Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025387.3 | NP_079663.1 | 2 - 3 | 204 | 71 | |
GenBank mRNA | AK003891.1 | - | 2 - 3 | 153 | 71 | |
AK008878.1 | - | 2 - 3 | 126 | 71 | ||
AK010609.1 | - | 2 - 3 | 164 | 71 | ||
AK012101.1 | - | 2 - 3 | 151 | 71 | ||
AK019395.1 | - | 2 - 3 | 153 | 71 | ||
AK050688.1 | - | 2 - 3 | 139 | 71 | ||
BC025854.1 | - | 2 - 3 | 105 | 5250659 | 71 | |
BC055862.1 | - | 2 - 3 | 128 | 3492090 | 71 | |
BC069899.1 | - | 1 - 2 | 850 | 71 | ||
BY785978.1 | - | 2 - 3 | 204 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |