Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022322.2 | NP_071717.1 | 3 - 4 | 521 | 145 | |
GenBank mRNA | AB055422.1 | - | 3 - 4 | 328 | 145 | |
AF191768.1 | - | 3 - 4 | 467 | 145 | ||
AF219993.1 | - | 3 - 4 | 394 | 145 | ||
AF291655.1 | - | 3 - 4 | 546 | 145 | ||
BC006919.1 | - | 3 - 4 | 330 | 3601223 | 145 | |
BC049944.1 | - | 3 - 4 | 521 | 2811628 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |