Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007922.5 | NP_031948.4 | 3 - 4 | 863 | 88 | |
GenBank mRNA | AK132354.1 | - | 3 - 4 | 856 | 88 | |
BC054474.1 | - | 3 - 4 | 847 | 30045506 | 88 | |
X87257.1 | - | 3 - 4 | 1133 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |