Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175150.3 | NP_780359.2 | 1 - 2 | 199 | 135 | |
GenBank mRNA | AK009878.1 | - | 1 - 2 | 127 | 135 | |
AK079517.1 | - | 1 - 2 | 146 | 135 | ||
AK160051.1 | - | 1 - 2 | 168 | 135 | ||
AK167701.1 | - | 1 - 2 | 107 | 135 | ||
AK167838.1 | - | 1 - 2 | 165 | 135 | ||
AK167977.1 | - | 1 - 2 | 164 | 135 | ||
BC062180.1 | - | 1 - 2 | 199 | 6336176 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |