Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009744.3 | NP_033874.1 | 4 - 5 | 474 | 88 | |
GenBank mRNA | AK036975.1 | - | 4 - 5 | 498 | 88 | |
AK039228.1 | - | 4 - 5 | 496 | 88 | ||
AK151229.1 | - | 4 - 5 | 499 | 88 | ||
BC052315.1 | - | 4 - 5 | 475 | 6309948 | 88 | |
D38377.1 | - | 4 - 5 | 463 | 88 | ||
U41465.1 | - | 3 - 4 | 486 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |