Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205321.1 | NP_001192250.1 | 15 - 16 | 2348 | 65 | |
NM_009134.3 | NP_033160.2 | 15 - 16 | 2348 | 65 | ||
GenBank mRNA | AK157866.1 | - | 15 - 16 | 2348 | 65 | |
AY538273.1 | - | 14 - 15 | 2278 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mus musculus Probe spans exons |