Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021496.3 | NP_067471.1 | 8 - 9 | 1551 | 93 | |
GenBank mRNA | AF195834.1 | - | 8 - 9 | 1358 | 93 | |
AK013863.1 | - | 636 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons |