Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172265.2 | NP_758469.1 | 9 - 10 | 1534 | 74 | |
GenBank mRNA | AK145801.1 | - | 9 - 10 | 1536 | 74 | |
AK160985.1 | - | 9 - 10 | 1463 | 74 | ||
BC027268.1 | - | 9 - 10 | 1481 | 74 | ||
BC038620.1 | - | 9 - 10 | 1534 | 4211776 | 74 | |
BC085255.1 | - | 9 - 10 | 1445 | 5342410 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |