Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028301.4 | NP_082577.2 | 12 - 13 | 1669 | 79 | |
GenBank mRNA | AK012494.1 | - | 12 - 13 | 1618 | 79 | |
AK129471.1 | - | 11 - 12 | 2698 | 79 | ||
BC022691.1 | - | 4 - 5 | 525 | 79 | ||
BC039644.1 | - | 5 - 6 | 622 | 79 | ||
BC050929.1 | - | 12 - 13 | 1605 | 6807194 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |