Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyzl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026915.2 | NP_081191.1 | 3 - 4 | 518 | 131 | |
GenBank mRNA | AK007412.1 | - | 487 | 131 | ||
AY572449.1 | - | 1 - 2 | 169 | 131 | ||
BC024932.1 | - | 3 - 4 | 518 | 3985345 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |