Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027704.2 | NP_081980.1 | 6 - 7 | 844 | 130 | |
GenBank mRNA | AK016865.1 | - | 6 - 7 | 855 | 130 | |
AY668948.1 | - | 5 - 6 | 785 | 130 | ||
BC116803.1 | - | 6 - 7 | 844 | 40126122 | 130 | |
BC116807.1 | - | 6 - 7 | 844 | 40126126 | 130 | |
BC144831.1 | - | 6 - 7 | 836 | 130 | ||
BC144832.1 | - | 6 - 7 | 844 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |