Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175523.4 | NP_780732.1 | 2 - 3 | 672 | 71 | |
GenBank mRNA | AK044610.1 | - | 2 - 3 | 742 | 71 | |
BC092238.1 | - | 2 - 3 | 672 | 30642218 | 71 | |
BC138473.1 | - | 1 - 2 | 478 | 8861493 | 71 | |
BC138474.1 | - | 1 - 2 | 478 | 8861494 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |