Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028958.4 | NP_083234.2 | 1 - 2 | 297 | 101 | |
GenBank mRNA | AF285574.1 | - | 1 - 2 | 101 | 101 | |
AK017109.1 | - | 1 - 2 | 298 | 101 | ||
BC106854.2 | - | 1 - 2 | 89 | 40046501 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |