Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bglap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AA016947.1 | - | 3 - 4 | 332 | 148 | |
AA475384.1 | - | 3 - 4 | 340 | 148 | ||
AK140913.1 | - | 5 - 6 | 1786 | 148 | ||
BE625312.1 | - | 3 - 4 | 337 | 148 | ||
BY414395.1 | - | 126 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Inventoried Probe spans exons |