Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019755.4 | NP_062729.1 | 2 - 3 | 292 | 107 | |
GenBank mRNA | AB031292.1 | - | 2 - 3 | 310 | 107 | |
AK003522.1 | - | 2 - 3 | 347 | 107 | ||
AK011282.1 | - | 2 - 3 | 346 | 107 | ||
AK154228.1 | - | 2 - 3 | 356 | 107 | ||
BC083078.1 | - | 2 - 3 | 335 | 6477560 | 107 | |
BC092095.1 | - | 2 - 3 | 311 | 3989985 | 107 | |
BC119088.1 | - | 2 - 3 | 292 | 8733837 | 107 | |
BC119090.1 | - | 2 - 3 | 292 | 8733839 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |