Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10608514 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608320 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608425 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608315 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608516 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608519 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608319 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608515 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608518 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608519 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608314 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608518 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608515 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608318 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608319 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608303 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608522 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608314 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608274 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608303 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608320 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608520 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608316 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608516 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608318 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608274 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608425 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608421 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608421 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608520 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608315 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608514 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608316 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10608522 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608317 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608424 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608517 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608521 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608420 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608420 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608317 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608273 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608313 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608513 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608517 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608302 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608424 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608313 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608513 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608521 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10608302 |
- |
- |
Mouse Genome 430 2.0 Array |
1449467_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1449467_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0Y00000235.mm.1 |
NM_009220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000235.mm.1 |
TC0Y00000034.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000230.mm.1 |
NM_001103152.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000230.mm.1 |
TC0Y00000034.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000231.mm.1 |
NM_001103152.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000231.mm.1 |
TC0Y00000034.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000232.mm.1 |
NM_001103152.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000232.mm.1 |
TC0Y00000034.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000236.mm.1 |
NM_009220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000236.mm.1 |
TC0Y00000034.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000229.mm.1 |
NM_001103152.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000229.mm.1 |
TC0Y00000034.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0Y00000234.mm.1 |
NM_009220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0Y00000234.mm.1 |
TC0Y00000034.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1449467_PM_at |
- |
- |
Mouse Expression Set 430 |
1449467_PM_at |
- |
- |
Mouse Expression Set 430 |
1449467_at |
- |
- |
Mouse Expression Set 430 |
1449467_at |
- |
- |
Murine Genome U74v2 Set |
161053_at |
- |
- |
Murine Genome U74v2 Set |
161053_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449467_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449467_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449467_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1449467_at |
- |
- |