Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033408.4 | NP_001028580.1 | 2 - 3 | 155 | 119 | |
GenBank mRNA | AK133355.1 | - | 2 - 3 | 156 | 119 | |
BC150950.1 | - | 2 - 3 | 117 | 9087861 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |