Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900010717.mm.1 |
NM_001037634.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010717.mm.1 |
NM_001037634.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010717.mm.1 |
TC1900001141.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010714.mm.1 |
NM_001037634.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010714.mm.1 |
NM_001037634.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010714.mm.1 |
TC1900001141.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010713.mm.1 |
NM_001037634.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010713.mm.1 |
NM_001037634.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010713.mm.1 |
TC1900001141.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010715.mm.1 |
NM_001037634.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010715.mm.1 |
NM_001037634.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010715.mm.1 |
TC1900001141.mm.1 |
MTA TC |