Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277216.1 | NP_001264145.1 | 7 - 8 | 749 | 146 | |
NM_001277217.1 | NP_001264146.1 | 7 - 8 | 775 | 146 | ||
NM_001277218.1 | NP_001264147.1 | 6 - 7 | 788 | 146 | ||
NM_001277220.1 | NP_001264149.1 | 7 - 8 | 923 | 146 | ||
NM_007560.4 | NP_031586.1 | 6 - 7 | 942 | 146 | ||
GenBank mRNA | AK086130.1 | - | 7 - 8 | 735 | 146 | |
AK140158.1 | - | 7 - 8 | 847 | 146 | ||
AK162844.1 | - | 6 - 7 | 623 | 146 | ||
BC065106.1 | - | 6 - 7 | 723 | 30357130 | 146 | |
BC065143.1 | - | 6 - 7 | 929 | 30629345 | 146 | |
BC138638.1 | - | 6 - 7 | 669 | 8861658 | 146 | |
BC138640.1 | - | 6 - 7 | 669 | 8861660 | 146 | |
Z23143.1 | - | 7 - 8 | 767 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |