Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253719.1 | NP_001240648.1 | 6 - 7 | 899 | 118 | |
NM_178673.4 | NP_848788.2 | 6 - 7 | 1159 | 118 | ||
GenBank mRNA | AF374460.1 | - | 6 - 7 | 1133 | 118 | |
AK033652.1 | - | 6 - 7 | 1150 | 118 | ||
AK046070.1 | - | 6 - 7 | 900 | 118 | ||
AK081152.1 | - | 6 - 7 | 1150 | 118 | ||
AK138388.1 | - | 6 - 7 | 1451 | 118 | ||
AK140611.1 | - | 6 - 7 | 1162 | 118 | ||
BC075640.1 | - | 6 - 7 | 1122 | 30545529 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |