Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sost | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024449.6 | NP_077769.4 | 2 | 640 | 137 | |
GenBank mRNA | AF326740.1 | - | 2 | 598 | 137 | |
AK017295.1 | - | 2 | 641 | 137 | ||
AK041382.1 | - | 2 | 641 | 137 | ||
BC137832.1 | - | 2 | 630 | 8860851 | 137 | |
BC137833.1 | - | 2 | 630 | 8860852 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |