Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026130.1 | NP_080406.1 | 2 - 3 | 285 | 107 | |
GenBank mRNA | AK004967.2 | - | 2 - 3 | 285 | 107 | |
AK007516.1 | - | 2 - 3 | 310 | 107 | ||
AK038017.1 | - | 2 - 3 | 273 | 107 | ||
BC021839.1 | - | 2 - 3 | 268 | 4207633 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |