Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400008665.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008665.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008665.mm.1 |
NM_177818.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008665.mm.1 |
NM_177818.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008665.mm.1 |
TC0400001193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008666.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008666.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008666.mm.1 |
NM_177818.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008666.mm.1 |
NM_177818.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008666.mm.1 |
TC0400001193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008668.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008668.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008668.mm.1 |
NM_177818.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008668.mm.1 |
NM_177818.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008668.mm.1 |
TC0400001193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008673.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008673.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008673.mm.1 |
TC0400001193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008674.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008674.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008674.mm.1 |
TC0400001193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008664.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008664.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008664.mm.1 |
NM_177818.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008664.mm.1 |
NM_177818.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008664.mm.1 |
TC0400001193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008667.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008667.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008667.mm.1 |
NM_177818.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008667.mm.1 |
NM_177818.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008667.mm.1 |
TC0400001193.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008678.mm.1 |
NM_001142775.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008678.mm.1 |
NM_001142775.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008678.mm.1 |
TC0400001193.mm.1 |
MTA TC |