Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007000.1 | - | 253 | 67 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | poliovirus receptor-related 2 | Pvrl2 |
AI325026, AI987993, Cd112, MPH, Pvr, Pvs, nectin-2 |
Anti-sense | Chr. 7 - 19716644 - 19749573 on NCBI Build GRCm38 | |
Downstream | basal cell adhesion molecule | Bcam |
1200005K12Rik, B-CAM, Gplu, Lu |
34.4kb | Anti-sense | Chr. 7 - 19756138 - 19770532 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |