Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Abhd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040172.1 | - | 225 | 66 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TAO kinase 1 | Taok1 |
2810468K05Rik, AU020252, D130018F14Rik, Map3k16, Markk, Psk2, mKIAA1361 |
Anti-sense | Chr. 11 - 77529162 - 77607860 on NCBI Build GRCm38 | |
Downstream | nuclear fragile X mental retardation protein interacting protein 2 | Nufip2 |
1110001M19Rik, 9530056D24Rik, AI256692, AI461731, PIG1, mKIAA1321 |
147.0kb | Sense | Chr. 11 - 77685650 - 77717966 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |