Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136459.1 | - | 1 | 1653 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FERM and PDZ domain containing 4 | Frmpd4 |
Gm196, PKAP1, Pdzd10, Pdzk10, Preso1, preso |
-74.0kb | Sense | Chr. X - 167471301 - 168577244 on NCBI Build GRCm38 |
Downstream | Rho GTPase activating protein 6 | Arhgap6 |
AI504284, RhoGAPX-1 |
141.1kb | Anti-sense | Chr. X - 168795094 - 169304440 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |