Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam160b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194345.1 | NP_919326.1 | 9 - 10 | 1199 | 77 | |
GenBank mRNA | AK090035.1 | - | 4 - 5 | 450 | 77 | |
BC050861.1 | - | 9 - 10 | 1200 | 77 | ||
CD348387.1 | - | 4 - 5 | 523 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |