Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019969.3 | NP_064353.2 | 3 - 4 | 381 | 104 | |
GenBank mRNA | AY574219.1 | - | 3 - 4 | 358 | 104 | |
BC139283.1 | - | 3 - 4 | 331 | 8862305 | 104 | |
BC139285.1 | - | 3 - 4 | 331 | 8862307 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |