Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133946.2 | NP_598707.2 | 6 - 7 | 2501 | 103 | |
GenBank mRNA | AK039531.1 | - | 3 - 4 | 1785 | 103 | |
AY355343.1 | - | 3 - 4 | 1439 | 103 | ||
BC013519.1 | - | 3 - 4 | 1408 | 103 | ||
BC031139.1 | - | 3 - 4 | 1493 | 4972485 | 103 | |
FJ222825.1 | - | 6 - 7 | 2447 | 103 | ||
JF810536.1 | - | 5 - 6 | 2886 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |