Search Thermo Fisher Scientific
Gene Symbol: Magea10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085506.1 | NP_001078975.1 | 3 - 4 | 243 | 108 | |
GenBank mRNA | AK136064.1 | - | 3 - 4 | 244 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |