Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027332.3 | NP_081608.1 | 2 - 3 | 315 | 112 | |
NM_027371.3 | NP_081647.2 | 2 - 3 | 315 | 112 | ||
GenBank mRNA | AK008988.1 | - | 2 - 3 | 304 | 112 | |
AK010079.1 | - | 2 - 3 | 312 | 112 | ||
AK162073.1 | - | 2 - 3 | 318 | 112 | ||
AK171035.1 | - | 2 - 3 | 296 | 112 | ||
AK171079.1 | - | 2 - 3 | 304 | 112 | ||
BC056165.1 | - | 2 - 3 | 266 | 5694049 | 112 | |
BC089369.1 | - | 2 - 3 | 300 | 30550759 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |