Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053079.2 | NP_444309.2 | 21 - 22 | 1862 | 82 | |
GenBank mRNA | AF205540.1 | - | 21 - 22 | 1861 | 82 | |
AK136428.1 | - | 21 - 22 | 1863 | 82 | ||
BC116248.1 | - | 21 - 22 | 1852 | 40093541 | 82 | |
BC116249.1 | - | 21 - 22 | 1852 | 40093542 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |