Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134185.2 | NP_598946.2 | 1 | 53 | 90 | |
GenBank mRNA | BC132241.1 | - | 1 | 53 | 40130518 | 90 |
BC132577.1 | - | 1 | 53 | 40130854 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |