Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177820.4 | NP_808488.2 | 1 - 2 | 130 | 112 | |
GenBank mRNA | AK033678.1 | - | 102 | 112 | ||
AK144113.1 | - | 1 - 2 | 133 | 112 | ||
BC147348.1 | - | 76 | 9056388 | 112 | ||
BC147349.1 | - | 76 | 9056408 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |