Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195075.1 | NP_001182004.1 | 9 - 10 | 915 | 76 | |
GenBank mRNA | CK780311.1 | - | 3 - 4 | 329 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |