Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110353.1 | 7 - 8 | 895 | 98 | ||
GenBank mRNA | AK030259.1 | - | 7 - 8 | 895 | 98 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1 | Riiad1 |
2310007A19Rik, Ncrna00166 |
-12.1kb | Anti-sense | Chr. 3 - 94464984 - 94475078 on NCBI Build GRCm38 |
Downstream | sorting nexin family member 27 | Snx27 |
5730552M22Rik, ESTM45, ESTM47, R75405 |
9.7kb | Anti-sense | Chr. 3 - 94497542 - 94582716 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |