Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppapdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121643.1 | 5 - 6 | 457 | 64 | ||
GenBank mRNA | BC089556.1 | - | 5 - 6 | 441 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Wolf-Hirschhorn syndrome candidate 1-like 1 (human) | Whsc1l1 |
6720429E03, A530023P05, AI528490, NSD3, WHISTLE |
-2.0kb | Sense | Chr. 8 - 25601175 - 25719667 on NCBI Build GRCm38 |
Downstream | DDHD domain containing 2 | Ddhd2 |
2010305K11Rik, SAMWD1, mKIAA0725 |
2.7kb | Anti-sense | Chr. 8 - 25725324 - 25754590 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |